Learn More
SLC6A3/DAT1 Antibody (CL3123) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46649IR
Product Code. 30291219
  • 737.00 EUR / 0.10mL
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC6A3/DAT1
Monoclonal
DyLight 755
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
This antibody was developed using a recombinant protein derived from Q01959, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
6531
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3123
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions