missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Antibody (CL2671) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46615MFV450
This item is not returnable.
View return policy
Description
NF-H Monoclonal antibody specifically detects NF-H in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| NF-H | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
| This antibody was developed using a recombinant protein derived from P12036, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 4744 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2671 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction