missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus M1R Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-30756JF669
This item is not returnable.
View return policy
Description
Monkeypox Virus M1R Polyclonal antibody specifically detects Monkeypox Virus M1R in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Monkeypox Virus M1R | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Monkeypox Virus M1R (RefSeq#: QNP13675.1; Ala3-Gly183) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| Janelia Fluor 669 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction