missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody (OTI7E10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-70881CL1
This item is not returnable.
View return policy
Description
Histone Deacetylase 2/HDAC2 Monoclonal antibody specifically detects Histone Deacetylase 2/HDAC2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Histone Deacetylase 2/HDAC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 415-488 of human HDAC2(NP_001518) produced in E.coli. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific, Stem Cell Markers | |
| 3066 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI7E10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction