missing translation for 'onlineSavingsMsg'
Learn More

GLOD4 Antibody [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30170550
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30170550 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30170550 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP318449JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLOD4 Polyclonal antibody specifically detects GLOD4 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLOD4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias C17orf25, CGI-150, chromosome 17 open reading frame 25, glyoxalase domain containing 4, glyoxalase domain-containing protein 4, HC71
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 263 and 313..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51031
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.