missing translation for 'onlineSavingsMsg'
Learn More

CPS1 Antibody (CPS1/8416), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21108PE

Product Code. 18669704

  • 702.00 EUR / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CPS1 Monoclonal antibody specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CPS1
Monoclonal
PE
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16
Recombinant human CPS1 protein
0.1 mL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
1373
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
CPS1/8416
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Canine
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.