missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axl Antibody (108724) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB154JF635
This item is not returnable.
View return policy
Description
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF,Gene Knock-Out
Specifications
| Axl | |
| Monoclonal | |
| Janelia Fluor 635 | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| Mouse myeloma cell line NS0-derived recombinant human Axl, Met1-Pro440, Accession # AAA61243 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF, Gene Knock-Out | |
| 108724 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 558 | |
| Store at 4°C in the dark. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur