Learn More
n-Propyl-2,2,3,3,3-d5-amine HCl, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Propylamine-D5 (propyl-2,2,3,3,3-D5) Hydrochloride |
| CAS | 1398065-66-1 |
| Molecular Formula | C3 D5 H4 N . Cl H |
| Quantity | 0.1 g |
| Synonym | 1-Propan-2,2,3,3,3-d5-amine hydrochloride, Propyl-2,2,3,3,3-d5-amine hydrochloride, n-Propyl-d5-amine hydrochloride, 1-Propylamine-2,2,3,3,3-d5 hydrochloride, Propylamine-D5 (propyl-2,2,3,3,3-D5) hydrochloride |
| InChI Formula | InChI=1 S/C3H9N.ClH/c1-2-3-4;/h2-4H2,1H3;1 H/i1D3,2D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])CN |
| IUPAC Name | 2,2,3,3,3-pentadeuteriopropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 100.602 |
| Formula Weight | 100.082 g/mol |
| Show More |