Learn More
L-Lysine-4,4,5,5-d4 HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Lysine-4,4,5,5-d4 Hydrochloride |
| CAS | 284664-96-6 |
| Molecular Formula | C6 2H4 H10 N2 O2 . Cl H |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1 H/t5-;/m0./s1/i1D2,2D2; |
| SMILES | Cl.[2 H]C([2 H])(CN)C([2 H])([2 H])C[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2,6-diamino-4,4,5,5-tetradeuteriohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 186.67 |
| Formula Weight | 186.1073 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |