Learn More
L-Lysine-3,3,4,4,5,5,6,6-d8 HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Lysine-3,3,4,4,5,5,6,6-d8 Hydrochloride |
| CAS | 344298-93-7 |
| Molecular Formula | C6 D8 H6 N2 O2 . Cl H |
| Quantity | 0.05 g |
| Synonym | L-Lysine-3,3,4,4,5,5,6,6-d8, hydrochloride (1:1) (ACI), L-Lysine-3,3,4,4,5,5,6,6-d8, monohydrochloride (9 CI), L-Lysine-3,3,4,4,5,5,6,6-D8 hydrochloride, L-Lysine-3,3,4,4,5,5,6,6-d8 hydrochloride, L-Lysine-d8 hydrochloride |
| InChI Formula | InChI=1 S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1 H/t5-;/m0./s1/i1D2,2D2,3D2,4D2; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2,6-diamino-3,3,4,4,5,5,6,6-octadeuteriohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 190.698 |
| Formula Weight | 190.132 g/mol |
| Show More |