Learn More
Di-n-decyl Phthalate-3,4,5,6-d4, CDN
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid Didecyl Ester D4 |
| CAS | 1276197-18-2 |
| Molecular Formula | C28 2H4 H42 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C28H46O4/c1-3-5-7-9-11-13-15-19-23-31-27(29)25-21-17-18-22-26(25)28(30)32-24-20-16-14-12-10-8-6-4-2/h17-18,21-22 H,3-16,19-20,23-24H2,1-2H3/i17D,18 D,21 D,22 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCCCC)c(C(=O)OCCCCCCCCCC)c1[2 H] |
| IUPAC Name | didecyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 450.6871 |
| Formula Weight | 450.3647 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |