Learn More
Nandrolone-16,16,17-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nandrolone-d3 |
| CAS | 120813-22-1 |
| Molecular Formula | C18 D3 H23 O2 |
| Quantity | 0.005 g |
| Synonym | Estr-4-en-3-one-16,16,17-d3, 17-hydroxy-, (17β)-, (17β)-17-Hydroxyestr-4-en-3-one-16,16,17-d3, Nandrolone-d3, d3-17β-Nandrolone, 19-Nortestosterone (16,16,17-D3), Nandrolone-16,16,17-D3 |
| InChI Formula | InChI=1 S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20 H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1/i7D2,17 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@@H]4 [C@H]3 CC[C@]2(C)[C@@]1([2 H])O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-17-hydroxy-13-methyl-2,6,7,8,9,10,11,12,14,15-decahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 277.416 |
| Formula Weight | 277.212 g/mol |
| Show More |