Learn More
Cymoxanil-d3 (methoxy-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cymoxanil methoxy-d3 |
| CAS | 2140803-92-3 |
| Molecular Formula | C7 2H3 H7 N4 O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C7H10N4O3/c1-3-9-7(13)10-6(12)5(4-8)11-14-2/h3H2,1-2H3,(H2,9,10,12,13)/b11-5+/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])O\N=C(/C#N)\C(=O)NC(=O)NCC |
| IUPAC Name | (1 E)-2-(ethylcarbamoylamino)-2-oxo-N-(trideuteriomethoxy)ethanimidoyl cyanide |
| Molecular Weight (g/mol) | 201.2 |
| Formula Weight | 201.0941 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |