Use promo code "25339" to get your promotional price.
p-Cymene, 99+%
Use promo code "25339" to get your promotional price.
| Quantity | 1 L |
|---|---|
| Packaging | Glass bottle |
??product.attributeSet.Chemical data??
| CAS | 99-87-6 |
|---|---|
| Molecular Formula | C10H14 |
| Molecular Weight (g/mol) | 134.22 |
| MDL Number | MFCD00008893 |
| InChI Key | HFPZCAJZSCWRBC-UHFFFAOYSA-N |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| PubChem CID | 7463 |
| ChEBI | CHEBI:28768 |
| IUPAC Name | 1-methyl-4-propan-2-ylbenzene |
| SMILES | CC1=CC=C(C=C1)C(C)C |
Specification
| Chemical Name or Material | p-Cymene |
|---|---|
| CAS | 99-87-6 |
| Melting Point | -68.0°C |
| Density | 0.8540g/mL |
| Boiling Point | 176.0°C to 178.0°C |
| Flash Point | 47°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C10H14 |
| Linear Formula | CH3C6H4CH(CH3)2 |
| Refractive Index | 1.4890 to 1.4910 |
| MDL Number | MFCD00008893 |
| Quantity | 1 L |
| Beilstein | 05, II, 322 |
| Merck Index | 15, 2760 |
| Specific Gravity | 0.854 |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Synonym | p-cymene,4-isopropyltoluene,p-isopropyltoluene,dolcymene,p-cymol,paracymene,cymene,para-cymene,1-isopropyl-4-methylbenzene,camphogen |
| Solubility Information | Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4 |
| InChI Key | HFPZCAJZSCWRBC-UHFFFAOYSA-N |
| SMILES | CC1=CC=C(C=C1)C(C)C |
| IUPAC Name | 1-methyl-4-propan-2-ylbenzene |
| Molecular Weight (g/mol) | 134.22 |
| PubChem CID | 7463 |
| ChEBI | CHEBI:28768 |
| Formula Weight | 134.22 |
| Percent Purity | 99+% |
| Physical Form | Liquid |
Technical documents
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=AQUA CHRONIC 2, sequence=2, description=LONG-TERM AQUATIC HAZARD Chronic 2}, {code=ASP. TOX. 1, sequence=3, description=Aspiration toxicity Category 1}, {code=FLAM. LIQ. 3, sequence=4, description=Flammable liquid Category 3}, {code=REPR. 2, sequence=5, description=Reproductive toxicity Category 2}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H304, sequence=2, description=May be fatal if swallowed and enters airways.}, {code=H331, sequence=3, description=Toxic if inhaled.}, {code=H361, sequence=4, description=Suspected of damaging fertility or the unborn child.}, {code=H411, sequence=5, description=Toxic to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P310, sequence=53, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P311, sequence=56, description=Call a POISON CENTER/doctor/ .}, {code=P331, sequence=57, description=Do NOT induce vomiting.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Reproductive toxicity Category 2
Hazard Statement
- H226- Flammable liquid and vapour.
- H304- May be fatal if swallowed and enters airways.
- H331- Toxic if inhaled.
- H361- Suspected of damaging fertility or the unborn child.
- H411- Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311- Call a POISON CENTER/doctor/ .
- P331- Do NOT induce vomiting.