Neodymium(III) nitrate hexahydrate, 99.9% (REO)
CAS: 16454-60-7 | H12N3NdO15 | 438.344 g/mol
Additional Details:
CAS Number:
16454-60-7
Weight:
0.05000kg
Transport:
UN number : 1477
Chem class : 5.1
Pack group : II
| Quantity | 50 g |
|---|
Disclaimers:
RUO – Research Use Only
??product.attributeSet.Chemical data??
| CAS | 16454-60-7 |
|---|---|
| Molecular Formula | H12N3NdO15 |
| Molecular Weight (g/mol) | 438.344 |
| MDL Number | MFCD00149799 |
| InChI Key | VQVDTKCSDUNYBO-UHFFFAOYSA-N |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| PubChem CID | 204494 |
| IUPAC Name | neodymium(3+);trinitrate;hexahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
Specification
| Chemical Name or Material | Neodymium(III) nitrate hexahydrate |
|---|---|
| Melting Point | ∼40°C |
| CAS | 16454-60-7 |
| pH | 3.0 to 4.0 |
| Physical Form | Crystalline |
| Quantity | 50 g |
| Assay Percent Notes | (REO) |
| Molecular Formula | H12N3NdO15 |
| MDL Number | MFCD00149799 |
| UN Number | UN1477 |
| Merck Index | 14,6450 |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Synonym | neodymium nitrate hexahydrate,neodymium trinitrate hexahydrate,neodymium nitrate monohydrate,neodymium iii nitrate, hexahydrate 1:3:6,nitric acid, neodymium 3+ salt, hexahydrate,ksc491s0r,neodymiumnitratehexahydrate,neodymium nitrate 6-hydrate,neodymium 3+ trinitrate hexahydrate,neodymium iii nitrate hexahydrate |
| Solubility Information | Soluble in water and alcohol. |
| InChI Key | VQVDTKCSDUNYBO-UHFFFAOYSA-N |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
| IUPAC Name | neodymium(3+);trinitrate;hexahydrate |
| Molecular Weight (g/mol) | 438.344 |
| PubChem CID | 204494 |
| Formula Weight | 438.34 (330.25 Anhydrous) |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
[Neodymium(III) nitrate hexahydrate]
[Danger]
[{code=H272, sequence=1, description=May intensify fire; oxidiser.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Neodymium(III) nitrate, hexahydrate}]
[{code=GHS03, sequence=1, description=FLAME OVER CIRCLE}]
Product Identifier
- Neodymium(III) nitrate hexahydrate
Signal Word
- Danger
Hazard Statement
- H272- May intensify fire; oxidiser.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental Information
- MIXTURE LIST- Contain: Neodymium(III) nitrate, hexahydrate