Boron trifluoride dihydrate, 65% BF3
CAS: 13319-75-0 | BF3H4O2 | 103.84 g/mol
Product Details
57.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: CAS Number: 13319-75-0 Weight: 0.16420kg Transport: UN number : 2851 Chem class : 8 Pack group : II
Quantity 100 mL
Packaging Glass bottle
Disclaimers: RUO – Research Use Only
Product Code. 10581875
Quantity Price
1 57.5  EUR / 100mL
Estimated Shipment
08-04-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
??product.attributeSet.Chemical data??
CAS 13319-75-0
Molecular Formula BF3H4O2
Molecular Weight (g/mol) 103.84
MDL Number MFCD00149901
InChI Key MJCYPBSRKLJZTB-UHFFFAOYSA-N
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
PubChem CID 61591
IUPAC Name trifluoroborane dihydrate
SMILES O.O.FB(F)F
Specification
Boiling Point 58°C to 60°C (1.5mmHg)
Chemical Name or Material Boron trifluoride dihydrate
Melting Point 6°C
CAS 13319-75-0
Color Colorless to Yellow
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 65.0 to 66%
Molecular Formula BF3H4O2
Linear Formula BF3·2H2O
MDL Number MFCD00149901
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Synonym boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
Solubility Information Solubility in water: miscible
InChI Key MJCYPBSRKLJZTB-UHFFFAOYSA-N
SMILES O.O.FB(F)F
IUPAC Name trifluoroborane dihydrate
Molecular Weight (g/mol) 103.84
PubChem CID 61591
Formula Weight 103.85
Percent Purity 65% (BF3)
Identification Confirmed
Packaging Glass bottle
Specific Gravity 1.642
Density 1.6420g/mL
Lewis acid