Three Step Guide to Selecting the Right Disposable Respirator
Filtering face piece (FFP) respirators, also called particulate respirators, dust masks or FFP masks, belong to the group known as air-purifying respirators and provide reliable protection against particulate pollutants such as dust, smoke and aerosols. As they are relatively lightweight, compact, easy to handle and require little maintenance, they are used for many common applications in sectors such as the automotive industry, agriculture, construction, pharmaceuticals, healthcare, chemicals and raw materials.
Disposable particulate respirators come with a wide range of protection levels, shapes and styles. Choosing the right respirator is key for optimal protection as respirators are designed for specific hazards. This guide helps you to select the disposable respiratory product that is the best fit for your application and desired comfort level in three steps.
You can find particulate respirators in three protection levels: FFP1, FFP2 and FFP3. These protection levels indicate the filter efficiency of the mask and are standardised across the European Union according to EN 149:2001+A1:2009.
FFP1
80%
… of all contaminants are filtered out of the air.
FFP1 masks protect against non-toxic water and oil-based particles at concentrations up to four times the workplace exposure limit (WEL). They are often used in environments with a low level of dust including from hand sanding, drilling and cutting.
FFP2
94%
… of all contaminants are filtered out of the air.
FFP2 masks protect against harmful water and oil-based particles at concentrations up to 10 times the workplace exposure limit. They are often used in applications with a moderate level of dust including from soft wood, metal, plastics (not PVC) and oil mist.
FFP3
99%
… of all contaminants are filtered out of the air.
FFP3 masks protect against harmful, carcinogenic and radioactive particles, as well as airborne biological agents of risk groups 2+3, at contaminant concentrations up to 20 times the workplace exposure limit. They are often used in applications with higher levels of dust and mists including from harmful pharmaceutical powders, biological agents and fibres.
Whether a mask is equipped with or without a valve has no influence on the level of protection. The exhalation valve increases comfort by reducing breathing effort and ensuring a more comfortable climate inside the mask. A valved mask is also less likely to fog up eyewear. On the other hand, unvalved masks offer lower unit cost and prevent wearers contaminating the environment.
Foldable masks help to save space when folded. However, unfolding and correct adjustment can be complicated and may lead to application errors. In contrast, cup-shaped masks are easy to handle, more resistant to deformation than folding masks and offer more comfort as well as a secure fit.
Standard ranges offer simple and effective protection for an affordable price, while premium ranges combine enhanced performance with increased comfort. Additional features of premium products include soft face piece sealing rings, valves, adjustable straps and more. Welding range respirators provide protection from mists, dusts, metal fumes and ozone during welding. There are also speciality respirators that are equipped with an active carbon filter for relief from odours in various applications, from welding to waste sorting.
Use our interactive selector tool to find your respirator you need with ease