accessibility menu, dialog, popup

UserName

CAS RN 96047-32-4

H3COOHF

CAS RN 96047-32-4

IUPAC Name: (3-fluoro-4-methoxyphenyl)methanol
Synonyms: (3-fluoro-4-methoxyphenyl)methanol
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: HHWYYUUOGAUCKX-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results