accessibility menu, dialog, popup

UserName

CAS RN 942206-06-6

ONBr

CAS RN 942206-06-6

IUPAC Name: 5-bromo-2-(furan-2-yl)pyridine
Synonyms: 5-bromo-2-(furan-2-yl)pyridine
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: ZKUDFBZRIQOMGO-UHFFFAOYSA-N
SMILES: BrC1=CN=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results