accessibility menu, dialog, popup

UserName

CAS RN 914636-99-0

FFSOOClF

CAS RN 914636-99-0

IUPAC Name: 2,3,5-trifluorobenzene-1-sulfonyl chloride
Synonyms: 2,3,5-trifluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 230.59
Molecular Formula: C6H2ClF3O2S
InChi Key: UOBDGEAGDFWWOQ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results