accessibility menu, dialog, popup

UserName

CAS RN 912761-79-6

NHOONH2

CAS RN 912761-79-6

IUPAC Name: 4-(piperidin-3-yloxy)benzamide
Synonyms: 4-(piperidin-3-yloxy)benzamide
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: OONWJLYEKQSBAT-UHFFFAOYNA-N
SMILES: NC(=O)C1=CC=C(OC2CCCNC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results