CAS RN 902837-45-0
CAS RN 902837-45-0
IUPAC Name:
3-iodo-2-propoxypyridine
Synonyms:
3-iodo-2-propoxypyridine
Molecular Weight (g/mol):
263.08
Molecular Formula:
C8H10INO
InChi Key:
WWMBXRKMRZDRFH-UHFFFAOYSA-N
SMILES:
CCCOC1=C(I)C=CC=N1
1
–
1
of
1
results
1
3-Iodo-2-n-propoxypyridine, 97%, Thermo Scientific™
CAS: 902837-45-0 Molecular Formula: C8H10INO Molecular Weight (g/mol): 263.078 MDL Number: MFCD07781164 InChI Key: WWMBXRKMRZDRFH-UHFFFAOYSA-N Synonym: 3-iodo-2-n-propoxypyridine,3-iodo-2-propoxy-pyridine PubChem CID: 42553132 IUPAC Name: 3-iodo-2-propoxypyridine SMILES: CCCOC1=C(C=CC=N1)I