accessibility menu, dialog, popup

UserName

CAS RN 902837-15-4

CH3HNONNNH

CAS RN 902837-15-4

IUPAC Name: N-methyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Synonyms: N-methyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 220.28
Molecular Formula: C11H16N4O
InChi Key: PKJNZUNKHVUUBC-UHFFFAOYSA-N
SMILES: CNC(=O)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results