accessibility menu, dialog, popup

UserName

H3CONOOOH2NCH3

CAS RN 9006-50-2

IUPAC Name: 5,5-dimethyl-2,4-dioxo-1,3-oxazolidine-3-carboxamide
Synonyms: albumen
Molecular Weight (g/mol): 172.14
Molecular Formula: C6H8N2O4
InChi Key: QCVGEOXPDFCNHA-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)N(C(N)=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results