accessibility menu, dialog, popup

UserName

CAS RN 886502-19-8

CH3ClNF

CAS RN 886502-19-8

IUPAC Name: 6-chloro-2-fluoro-3-methylbenzonitrile
Synonyms: 6-chloro-2-fluoro-3-methylbenzonitrile
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H5ClFN
InChi Key: ZZIYYBYHLBGXCD-UHFFFAOYSA-N
SMILES: CC1=C(F)C(C#N)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results