accessibility menu, dialog, popup

UserName

CAS RN 885268-00-8

CH3ONH2OHO

CAS RN 885268-00-8

IUPAC Name: 2-amino-5-(2-methylphenoxy)benzoic acid
Synonyms: 2-amino-5-(2-methylphenoxy)benzoic acid
Molecular Weight (g/mol): 243.26
Molecular Formula: C14H13NO3
InChi Key: XJZGSKLLSIYBQF-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC1=CC(C(O)=O)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results