accessibility menu, dialog, popup

UserName

CAS RN 879904-88-8

CH3SiH3COONa

CAS RN 879904-88-8

IUPAC Name: sodium (furan-2-yl)dimethylsilanolate
Synonyms: sodium furan-2-yldimethylsilanolate
Molecular Weight (g/mol): 164.21
Molecular Formula: C6H9NaO2Si
InChi Key: LZJRCLPTAZMQQW-UHFFFAOYSA-N
SMILES: [Na+].C[Si](C)([O-])C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results