CAS RN 874287-99-7
CAS RN 874287-99-7
IUPAC Name:
{4-[(4-bromophenyl)carbamoyl]phenyl}boronic acid
Synonyms:
4-[(4-bromophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
319.95
Molecular Formula:
C13H11BBrNO3
InChi Key:
FBTLHSKBTDKVGQ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(C=C1)C(=O)NC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4-(4-Bromophenylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™
CAS: 874287-99-7 Molecular Formula: C13H11BBrNO3 Molecular Weight (g/mol): 319.949 MDL Number: MFCD09027201 InChI Key: FBTLHSKBTDKVGQ-UHFFFAOYSA-N Synonym: n-4-bromophenyl 4-boronobenzamide,4-4-bromophenyl carbamoyl phenyl boronic acid,4-4-bromophenylcarbamoyl phenylboronic acid,4-4-bromophenyl carbamoyl phenylboronic acid,4-4-bromophenyl carbamoyl benzeneboronic acid,boronic acid,b-4-4-bromophenyl amino carbonyl phenyl,acmc-209qlo,n4-bromophenyl 4-boronobenzamide,4-4-bromophenyl amino carbonyl phenylboronic acid PubChem CID: 44119160 IUPAC Name: [4-[(4-bromophenyl)carbamoyl]phenyl]boronic acid SMILES: B(C1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)Br)(O)O