accessibility menu, dialog, popup

UserName

CAS RN 870849-56-2

H3CFOCH3OH

CAS RN 870849-56-2

IUPAC Name: 1-(4-fluoro-3-methoxyphenyl)ethan-1-ol
Synonyms: 1-(4-fluoro-3-methoxyphenyl)ethanol
Molecular Weight (g/mol): 170.18
Molecular Formula: C9H11FO2
InChi Key: OGBGGCJKDXCBGP-UHFFFAOYNA-N
SMILES: COC1=C(F)C=CC(=C1)C(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results