accessibility menu, dialog, popup

UserName

CAS RN 850568-50-2

BOOCH3HOOH

CAS RN 850568-50-2

IUPAC Name: [3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
Synonyms: 3-(2-methyl-1,3-dioxolan-2-yl)phenylboronic acid
Molecular Weight (g/mol): 208.02
Molecular Formula: C10H13BO4
InChi Key: NSDRSKQWUQJUAE-UHFFFAOYSA-N
SMILES: CC1(OCCO1)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results