CAS RN 82386-89-8
CAS RN 82386-89-8
IUPAC Name:
2-chloro-5-(trifluoromethyl)benzaldehyde
Synonyms:
2-chloro-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
208.56
Molecular Formula:
C8H4ClF3O
InChi Key:
OZZOJJJYKYKBNH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(Cl)C(C=O)=C1
1
–
1
of
1
results
1
2-Chloro-5-(trifluoromethyl)benzaldehyde, 97+%, Thermo Scientific™
CAS: 82386-89-8 Molecular Formula: C8H4ClF3O Molecular Weight (g/mol): 208.564 MDL Number: MFCD00134430 InChI Key: OZZOJJJYKYKBNH-UHFFFAOYSA-N Synonym: 2-chloro-5-trifluoromethyl benzaldehyde,benzaldehyde, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl-benzaldehyde,pubchem2881,acmc-209pop,ksc495k9f,timtec-bb sbb003466,4-chloro-3-formylbenzotrifluoride PubChem CID: 688181 IUPAC Name: 2-chloro-5-(trifluoromethyl)benzaldehyde SMILES: C1=CC(=C(C=C1C(F)(F)F)C=O)Cl