CAS RN 797791-33-4
CAS RN 797791-33-4
IUPAC Name:
6-chloro-2,3-difluorobenzaldehyde
Synonyms:
6-chloro-2,3-difluorobenzaldehyde
Molecular Weight (g/mol):
176.55
Molecular Formula:
C7H3ClF2O
InChi Key:
YVKZXQCYLHFHIR-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(C=O)=C(Cl)C=C1
1
–
1
of
1
results
1
6-Chloro-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™
CAS: 797791-33-4 Molecular Formula: C7H3ClF2O Molecular Weight (g/mol): 176.547 MDL Number: MFCD07368118 InChI Key: YVKZXQCYLHFHIR-UHFFFAOYSA-N PubChem CID: 9812909 IUPAC Name: 6-chloro-2,3-difluorobenzaldehyde SMILES: C1=CC(=C(C(=C1F)F)C=O)Cl