accessibility menu, dialog, popup

UserName

CAS RN 768-70-7

CH3OHC

CAS RN 768-70-7

IUPAC Name: 1-ethynyl-3-methoxybenzene
Synonyms: 1-ethynyl-3-methoxybenzene
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: ZASXCTCNZKFDTP-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C#C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)