accessibility menu, dialog, popup

UserName

CAS RN 76265-69-5

OONH(S)NHOFFFOOH

CAS RN 76265-69-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-(2,2,2-trifluoroacetamido)hexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-(2,2,2-trifluoroacetamido)hexanoic acid
Molecular Weight (g/mol): 464.44
Molecular Formula: C23H23F3N2O5
InChi Key: ZVLMWTPNDXNXSZ-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC(=O)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)