accessibility menu, dialog, popup

UserName

CAS RN 7522-43-2

CH3H3C(S)NH3OHOO

CAS RN 7522-43-2

IUPAC Name: (2S)-2-azaniumyl-2-(hydroxymethyl)-3-methylbutanoate
Synonyms: (2S)-2-ammonio-2-(hydroxymethyl)-3-methylbutanoate
Molecular Weight (g/mol): 147.17
Molecular Formula: C6H13NO3
InChi Key: QTYSAGIKKRDGIV-ZCFIWIBFSA-N
SMILES: CC(C)[C@]([NH3+])(CO)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results