accessibility menu, dialog, popup

UserName

CAS RN 75039-84-8

H3COH(E)

CAS RN 75039-84-8

IUPAC Name: (2E)-undec-2-en-1-ol
Synonyms: 2-undecenol
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: SKBIQKQBLQHOSU-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\CO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results