accessibility menu, dialog, popup

UserName

CAS RN 74461-64-6

ClNSClN

CAS RN 74461-64-6

IUPAC Name: 5-chloro-3-(chloromethyl)-1,2,4-thiadiazole
Synonyms: 5-chloro-3-(chloromethyl)-1,2,4-thiadiazole
Molecular Weight (g/mol): 169.02
Molecular Formula: C3H2Cl2N2S
InChi Key: FFFMIFUBJZBTCG-UHFFFAOYSA-N
SMILES: ClCC1=NSC(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results