CAS RN 73647-09-3
CAS RN 73647-09-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Guanosine 2’,3’-Cyclic Monophosphate Triethylamine Salt, TRC
CAS: 73647-09-3 Molecular Formula: C10 H12 N5 O7 P . C6 H15 N Molecular Weight (g/mol): 446.395 Synonym: Guanosine Cyclic 2',3'-(Hydrogen Phosphate) N,N-Diethylethanamine; IUPAC Name: 9-[(3aR,4R,6R,6aR)-2-hydroxy-6-(hydroxymethyl)-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-4-yl]-2-amino-3H-purin-6-one;N,N-diethylethanamine SMILES: CCN(CC)CC.NC1=NC(=O)c2ncn([C@@H]3O[C@H](CO)[C@H]4OP(=O)(O)O[C@@H]34)c2N1