accessibility menu, dialog, popup

UserName

CAS RN 7252-83-7

CH3OBrOH3C

CAS RN 7252-83-7

IUPAC Name: 2-bromo-1,1-dimethoxyethane
Synonyms: ethane, 2-bromo-1,1-dimethoxy-
Molecular Weight (g/mol): 169.02
Molecular Formula: C4H9BrO2
InChi Key: FUSFWUFSEJXMRQ-UHFFFAOYSA-N
SMILES: COC(CBr)OC
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)