CAS RN 725-14-4
CAS RN 725-14-4
IUPAC Name:
4'-methoxy-[1,1'-biphenyl]-4-carboxylate
Synonyms:
4'-methoxy-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol):
227.24
Molecular Formula:
C14H11O3
InChi Key:
FDPKGXQCDURRBM-UHFFFAOYSA-M
SMILES:
COC1=CC=C(C=C1)C1=CC=C(C=C1)C([O-])=O
1
–
1
of
1
results
1
4'-Methoxybiphenyl-4-carboxylic acid, 95%, Thermo Scientific™
CAS: 725-14-4 Molecular Formula: C14H12O3 Molecular Weight (g/mol): 228.247 MDL Number: MFCD00185792 InChI Key: FDPKGXQCDURRBM-UHFFFAOYSA-N Synonym: 4'-methoxybiphenyl-4-carboxylic acid,4-4-methoxyphenyl benzoic acid,4'-methoxy-1,1'-biphenyl-4-carboxylic acid,4'-methoxy 1,1'-biphenyl-4-carboxylic acid,4'-methoxy-biphenyl-4-carboxylic acid,4'-methoxy-4-biphenylcarboxylic acid,4-biphenyl-4'-methoxy carboxylic acid,1,1'-biphenyl-4-carboxylicacid, 4'-methoxy,pubchem10293,acmc-1bc5q PubChem CID: 2759552 IUPAC Name: 4-(4-methoxyphenyl)benzoic acid SMILES: COC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O