accessibility menu, dialog, popup

UserName

CAS RN 71-27-2

ClClH3CNCH3H3COOOONCH3CH3H3C

CAS RN 71-27-2

IUPAC Name: trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dichloride
Synonyms: succinylcholine chloride bis(succinyldichlorocholine) succinylcholine dichloride (anhydrous) anhydrous succinylcholine chloride suxamethonium chloride (anhydrous) chlorsuccinylcholin suxamethonium dichloride anhydrous succinyldicholine dichloride succinyldicholine dichloride (anhydrous) anhydrous suxamethonium chloride
Molecular Weight (g/mol): 361.3
Molecular Formula: C14H30Cl2N2O4
InChi Key: YOEWQQVKRJEPAE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C

 No Refinements