CAS RN 704869-38-5
CAS RN 704869-38-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SUN11602, MedChemExpress
MedChemExpress SUN11602 is a novel aniline compound with basic fibroblast growth factor-like activity.