accessibility menu, dialog, popup

UserName

CAS RN 6935-27-9

NHN

CAS RN 6935-27-9

IUPAC Name: N-benzylpyridin-2-amine
Synonyms: N-benzylpyridin-2-amine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: WYHXNQXDQQMTQI-UHFFFAOYSA-N
SMILES: C(NC1=CC=CC=N1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)