accessibility menu, dialog, popup

UserName

CAS RN 69123-98-4

HOOFHONIOHNO

CAS RN 69123-98-4

IUPAC Name: 1-[3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: fialuridine
Molecular Weight (g/mol): 372.09
Molecular Formula: C9H10FIN2O5
InChi Key: IPVFGAYTKQKGBM-UHFFFAOYNA-N
SMILES: OCC1OC(C(F)C1O)N1C=C(I)C(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results