accessibility menu, dialog, popup

UserName

CAS RN 68443-63-0

H3COOCH3CH3

CAS RN 68443-63-0

IUPAC Name: butyl 2-ethylhexanoate
Synonyms: butyl 2-ethylhexanoate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: USPADFUBVAGYOJ-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C(CC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results