CAS RN 68359-37-5
CAS RN 68359-37-5
IUPAC Name:
cyano(4-fluoro-3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms:
β-cyfluthrin cyfluthrine epa pesticide chemical code 128831 tempo 2 eulan SP cylence cyfoxylate solfac cyfluthine cyfluthrin
Molecular Weight (g/mol):
434.29
Molecular Formula:
C22H18Cl2FNO3
InChi Key:
QQODLKZGRKWIFG-UHFFFAOYNA-N
SMILES:
CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)C1=CC=C(F)C(OC2=CC=CC=C2)=C1
1
–
6
of
6
results
2
Cyfluthrin, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality