CAS RN 681435-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 681435-25-6 Molecular Weight (g/mol): 272.22 MDL Number: MFCD00150204 InChI Key: KPWJBEFBFLRCLH-UHFFFAOYSA-L Synonym: cadmium 2+ dibromide,cadmium 2+ ion dibromide,cadmium bromide, anhydrous 25g,cadmium bromide, anhydrous, IUPAC Name: cadmium(2+) dibromide SMILES: [Br-].[Br-].[Cd++]