accessibility menu, dialog, popup

UserName

CAS RN 675602-92-3

NFFFOOH

CAS RN 675602-92-3

IUPAC Name: 2,3,6-trifluoropyridine-4-carboxylic acid
Synonyms: 2,3,6-trifluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 177.08
Molecular Formula: C6H2F3NO2
InChi Key: RXNORRUCMWSIQZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=NC(F)=C1F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results