accessibility menu, dialog, popup

UserName

CAS RN 67392-87-4

H3C(R)O(R)(R)(R)(S)(S)H3C(S)(S)(S)(S)OO

CAS RN 67392-87-4

IUPAC Name: (1R,2R,4R,10R,11S,14S,15S,16S,18S,19S)-10,14-dimethylspiro[hexacyclo[9.8.0.0²,⁴.0⁵,¹⁰.0¹⁴,¹⁹.0¹⁶,¹⁸]nonadecane-15,2'-oxolan]-5-ene-5',7-dione
Synonyms: yasmin drospirenone DRSP dihydrospirorenone drospirenona drospirenonum 1,2-dihydrospirorenone drospirenona [inn-spanish] drospirenone (product) dehydrospirorenone
Molecular Weight (g/mol): 366.5
Molecular Formula: C24H30O3
InChi Key: METQSPRSQINEEU-HXCATZOESA-N
SMILES: C[C@]12CC[C@H]3[C@@H]([C@H]4C[C@H]4C4=CC(=O)CC[C@]34C)[C@@H]1[C@@H]1C[C@@H]1[C@@]21CCC(=O)O1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results