accessibility menu, dialog, popup

UserName

CAS RN 66644-79-9

CH3NHSH2NBr

CAS RN 66644-79-9

IUPAC Name: (2-bromo-4-methylphenyl)thiourea
Synonyms: 2-bromo-4-methylphenylthiourea
Molecular Weight (g/mol): 245.14
Molecular Formula: C8H9BrN2S
InChi Key: QBTARKBNALVKLA-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(NC(N)=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results